Learn More
Eurofins Discoverx Raf-1 (truncated), active, 10 µg. (14-352)
Supplier:  Eurofins Discoverx 14352
Encompass
N-terminal GST-tagged recombinant, human Raf-1 amino acids 306-end, containing the mutations Y340D and Y341D. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8783