Learn More
Medchemexpress LLC Raf inhibitor 1 | 1093100-40-3 | 98.0% | C26H19ClN8 | 50 MG
SDP
Catalog No. 5000356966
Encompass_Preferred
Raf inhibitor 1 is a potent Raf kinase inhibitor with Ki values of 1 nM, 1 nM, and 0.3 nM for B-RafWT, B-RafV600E, and C-Raf, respectively. It inhibits A375 and HCT-116 proliferation with IC50s of 0.31 and 0.72 μM. This inhibitor binds to and stabilizes B-Raf in a DFG-out, inactive conformation where the ATP pocket is partially filled by Phe595 and Gly596 from the DFG motif. It also shows low micromolar inhibition against wild type B-Raf cell lines, which may be due to off-target kinase activities or pan-Raf inhibition, including Raf dimers. This product is for research use only.
  • Potent Raf kinase inhibitor
  • Inhibits B-RafWT, B-RafV600E, and C-Raf
  • Inhibits A375 and HCT-116 proliferation
  • Binds to and stabilizes B-Raf in an inactive conformation
  • Suitable for research applications
Catalog No. 50-003-56966 Supplier Medchemexpress LLC Supplier No. HY1417750MG
Raf inhibitor 1 is a potent Raf kinase inhibitor with Ki values of 1 nM, 1 nM, and 0.3 nM for B-RafWT, B-RafV600E, and C-Raf, respectively. It inhibits A375 and HCT-116 proliferation with IC50s of 0.31 and 0.72 μM. This inhibitor binds to and stabilizes B-Raf in a DFG-out, inactive conformation where the ATP pocket is partially filled by Phe595 and Gly596 from the DFG motif. It also shows low micromolar inhibition against wild type B-Raf cell lines, which may be due to off-target kinase activities or pan-Raf inhibition, including Raf dimers. This product is for research use only.
  • Potent Raf kinase inhibitor
  • Inhibits B-RafWT, B-RafV600E, and C-Raf
  • Inhibits A375 and HCT-116 proliferation
  • Binds to and stabilizes B-Raf in an inactive conformation
  • Suitable for research applications