Learn More
Carnabio Usa Inc RAF1/5UG/CATALYTIC
Supplier:  Carnabio Usa Inc 091255
Encompass
RAF1, Catalytic domain, Active mutant [Y340D], [Y341D], Amino Acid 306-648, NP_002871.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0603