Learn More
Medchemexpress LLC Raf1 protein, human (Y340D, Y341D, sf9, His, GST) | ≥80.0% | 10UG
SDP
Catalog No. 5000175151
Encompass_Preferred
Recombinant human RAF1 protein containing Y340D and Y341D activating mutations. Produced in Sf9 insect cells and expressed with N-terminal His and GST tags, this kinase fragment (residues S306-F648) is supplied as a filtered solution formulated in 50 mM HEPES pH 7.5, 200 mM NaCl, 20% glycerol, and 1 mM DTT. It is designed for biochemical assays and studies of MAPK/ERK signaling.
  • Activating Y340D and Y341D mutations enhance kinase activity.
  • Expressed in Sf9 insect cells for eukaryotic folding and modifications.
  • N-terminal His and GST tags facilitate purification and detection.
  • Supplied as a ready-to-use filtered solution; no reconstitution required.
  • Formulated for stability with 20% glycerol and a reducing agent.
  • Suitable for kinase assays, signaling studies, and protein-protein interaction experiments.
  • Store at -80°C and avoid repeated freeze-thaw cycles.
Catalog No. 50-001-75151 Supplier Medchemexpress LLC Supplier No. HYP70273310UG
Recombinant human RAF1 protein containing Y340D and Y341D activating mutations. Produced in Sf9 insect cells and expressed with N-terminal His and GST tags, this kinase fragment (residues S306-F648) is supplied as a filtered solution formulated in 50 mM HEPES pH 7.5, 200 mM NaCl, 20% glycerol, and 1 mM DTT. It is designed for biochemical assays and studies of MAPK/ERK signaling.
  • Activating Y340D and Y341D mutations enhance kinase activity.
  • Expressed in Sf9 insect cells for eukaryotic folding and modifications.
  • N-terminal His and GST tags facilitate purification and detection.
  • Supplied as a ready-to-use filtered solution; no reconstitution required.
  • Formulated for stability with 20% glycerol and a reducing agent.
  • Suitable for kinase assays, signaling studies, and protein-protein interaction experiments.
  • Store at -80°C and avoid repeated freeze-thaw cycles.