Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Raf265 (CHIR-265) | 927880-90-8 | MFCD16659061 | 99.8% | 518.41 g/mol | C24H16F6N6O | 25 MG
SDP

Catalog No. 5000245863
Encompass_Preferred

RAF265 (CHIR-265) is a small-molecule, orally active RAF and VEGFR2 kinase inhibitor used in oncology research and preclinical studies.

  • Potent RAF and VEGFR2 inhibitor for research applications.
  • Orally active small molecule.
  • Molecular formula C24H16F6N6O.
  • Molecular weight 518.41 g/mol.
  • Physical form: powder.
  • Purity: 99.8%.
  • Storage: powder -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-45863 Supplier Medchemexpress LLC Supplier No. HY1024825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RAF265 (CHIR-265) is a small-molecule, orally active RAF and VEGFR2 kinase inhibitor used in oncology research and preclinical studies.

  • Potent RAF and VEGFR2 inhibitor for research applications.
  • Orally active small molecule.
  • Molecular formula C24H16F6N6O.
  • Molecular weight 518.41 g/mol.
  • Physical form: powder.
  • Purity: 99.8%.
  • Storage: powder -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.