Learn More
TARGETMOL CHEMICALS INC RAF709 10MG
SDP
Catalog No. 502576879
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RAF709 is a novel Raf kinase inhibitor with IC50s of 0.5 and 1.8 nM for c-Raf and b-Raf respectively. purity: 99%
Catalog No. 50-257-6879 Supplier TARGETMOL CHEMICALS INC Supplier No. T371110MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RAF709 is a novel Raf kinase inhibitor with IC50s of 0.5 and 1.8 nM for c-Raf and b-Raf respectively. purity: 99%