Learn More
TARGETMOL CHEMICALS INC RAFABEGRON 5MG
SDP
Catalog No. 502580273
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Rafabegron (TAK677) is a potent and selective (beta)3 adrenergic receptor agonist for the study of diabetes and obesity. purity: 100%
Catalog No. 50-258-0273 Supplier TARGETMOL CHEMICALS INC Supplier No. T342565MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Rafabegron (TAK677) is a potent and selective (beta)3 adrenergic receptor agonist for the study of diabetes and obesity. purity: 100%