Learn More
Selleck Chemical LLC Raffinose S5155-25mg
SDP
Catalog No. 503108506
Encompass_Preferred
Raffinose (Melitose Melitriose) is a complex carbohydrate a trisaccharide composed of galactose fructose and glucose It can be broken apart into galactose and sucrose via the enzyme alpha-galactosidase
Catalog No. 50-310-8506 Supplier Selleck Chemical LLC Supplier No. S515525MG
Raffinose (Melitose Melitriose) is a complex carbohydrate a trisaccharide composed of galactose fructose and glucose It can be broken apart into galactose and sucrose via the enzyme alpha-galactosidase