Learn More
AG Scientific Inc Raffinose Pentahydrate 25 G
Catalog No. 502268372
Encompass_Preferred
Raffinose pentahydrate is a trisaccharide composed of galactose, fructose, and glucose. It is hydrolyzed to D-galactose and sucrose by the actions of D-galactosidase. Raffinose has applications in cryopreservation, where it is used to provide hypertonicity for cell desiccation before freezing.
Catalog No. 50-226-8372 Supplier AG Scientific Inc Supplier No. R262125GM
Raffinose pentahydrate is a trisaccharide composed of galactose, fructose, and glucose. It is hydrolyzed to D-galactose and sucrose by the actions of D-galactosidase. Raffinose has applications in cryopreservation, where it is used to provide hypertonicity for cell desiccation before freezing.