Learn More
Medchemexpress LLC Raffinose (Standard) | 512-69-6 | 10 MG
SDP
Catalog No. 5000418984
Encompass_Preferred
Raffinose (Standard) is an analytical standard intended for research and analytical applications. It regulates intestinal flora, inhibits the TLR4-MyD88-NF-κB signaling pathway, and activates the Nrf2 signaling pathway. Raffinose exhibits anti-inflammatory, antioxidant, and immunomodulatory activities, and is orally active. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard of Raffinose
  • Intended for research and analytical applications
  • Regulates intestinal flora
  • Inhibits the TLR4-MyD88-NF-κB signaling pathway
  • Activates the Nrf2 signaling pathway
  • Exhibits anti-inflammatory activity
  • Exhibits antioxidant activity
  • Exhibits immunomodulatory activities
  • Orally active
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-18984 Supplier Medchemexpress LLC Supplier No. HYN7088R10MG
Raffinose (Standard) is an analytical standard intended for research and analytical applications. It regulates intestinal flora, inhibits the TLR4-MyD88-NF-κB signaling pathway, and activates the Nrf2 signaling pathway. Raffinose exhibits anti-inflammatory, antioxidant, and immunomodulatory activities, and is orally active. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard of Raffinose
  • Intended for research and analytical applications
  • Regulates intestinal flora
  • Inhibits the TLR4-MyD88-NF-κB signaling pathway
  • Activates the Nrf2 signaling pathway
  • Exhibits anti-inflammatory activity
  • Exhibits antioxidant activity
  • Exhibits immunomodulatory activities
  • Orally active
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS