Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rafoxanide 98.0+%, TCI America™
SDP

Catalog No. R01081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R01081G Supplier TCI America Supplier No. R01081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22662-39-1
Molecular Formula C19H11Cl2I2NO3
Molecular Weight (g/mol) 626.01
MDL Number MFCD00682899
InChI Key NEMNPWINWMHUMR-UHFFFAOYSA-N
Synonym 3′C-Chloro-4′C-(4-chlorophenoxy)-3,5-diiodosalicylanilide
PubChem CID 31475
IUPAC Name N-[3-chloro-4-(4-chlorophenoxy)phenyl]-2-hydroxy-3,5-diiodobenzamide
SMILES OC1=C(I)C=C(I)C=C1C(=O)NC1=CC=C(OC2=CC=C(Cl)C=C2)C(Cl)=C1

Specifications

Melting Point 175°C
Quantity 1 g
Formula Weight 626.01
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Rafoxanide
RTECSNumber VN7870000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.