Learn More
Aviva Systems Biology Corp Rag1 Antibody - C-terminal region (ARP42949_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42949_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rag1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Rag-1ApplicationWB. Species reactivity: Horse, Mouse, Pig, Rat
Catalog No. 50-285-047