Learn More
ABclonal Technology RAG1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1264620UL
Encompass_Preferred
The protein encoded by this gene is involved in activation of immunoglobulin V-D-J recombination. The encoded protein is involved in recognition of the DNA substrate, but stable binding and cleavage activity also requires RAG2. Defects in this gene can be the cause of several diseases.
Catalog No. 50-000-99364