Learn More
Aviva Systems Biology Corp RAGE Antibody - middle region (ARP56744_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56744_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAGE. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RAGE. Alternate names: MOK, RAGE1, RAGE, RAGE-1ApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-286-206