Learn More
Stoelting™ Ragnell Retractor
SDP
Catalog No. 10001425 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
3 x 5, 5 x 15
Catalog No. Model
10-001-425 3 x 5, 5 x 15
1 options
Catalog No. 10-001-425 Supplier Stoelting™ Supplier No. 5212624
Add to Cart
Add to Cart
Stoelting™ Ragnell Retractors can be used to actively separate the edges of a surgical incision or wound or hold back underlying organs and tissues so that body parts under the incision may be accessed.
  • Sharp retractors are better at providing a secure grip and they can safely hold back the skin, muscles, fascia, tendons, or the periosteum but they may damage vessels, nerves, the peritoneum, glands or cysts
  • Blunt retractors are recommended when iatrogenic injury is a concern.
  • The main benefit of blunt retractors is that they prevent unintended injury to the tissue held by the retractor or to the underlying tissue.
Specifications
For Use With (Application) Neuroscience
Length (English) 6 in.
Model 3 x 5, 5 x 15