Learn More
Aviva Systems Biology Corp RAI14 Antibody - middle region (ARP55286_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55286_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAI14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RAI14. Alternate names: DKFZp564G013, KIAA1334, NORPEG, RAI13ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61456