Learn More
Spherotech RAINBOW CAL PART 5ML 3-3.4
Catalog No. NC9236843
Encompass_Preferred
Rainbow Calibration Particles, 6 peaks, 1E7/mL, 3.0-3.4 um, 5 mL
Catalog No. NC9236843 Supplier Spherotech Supplier No. RCP305
Rainbow Calibration Particles, 6 peaks, 1E7/mL, 3.0-3.4 um, 5 mL