Learn More
Spherotech Rainbow Calibration Particles, 8 peaks 107/mL, 3.0-3.4 µm, 5ML
Supplier:  Spherotech RCP305A
Encompass_Preferred
The SPHERO™ Calibration Particles are designed for routine calibration of flow cytometers. By using these particles, the sensitivity and linearity of the system can be determined. The sensitivity of the instrument will help detect any dye or light contamination. The linearity of the instrument will determine if the logarithmic amps and PMTs are functioning properly.
Catalog No. NC9572794