Learn More
Spherotech Rainbow Calibration Particles, 8 peaks, 1E7/mL, 3.0-3.4µm, 20mL
Catalog No. NC0955335
Encompass_Preferred
Rainbow Calibration Particles, 8 peaks, 1E7/mL, 3.0-3.4 um, 20 mL
Catalog No. NC0955335 Supplier Spherotech Supplier No. RCP3020A
Rainbow Calibration Particles, 8 peaks, 1E7/mL, 3.0-3.4 um, 20 mL