Learn More
Andwin Scientific RAINBOW CRYOTAGS 1.5X.75
SDP
Supplier:  Andwin Scientific RNBW2100
Encompass_Preferred
The Laser Cryo-Babies measure 50 x 0.75". This product features compatibility with laser printers as the labels are heat-resistant and have a stay-flat design. The labels can withstand temperatures up to 150C and can edure boiling water baths and freezer temperatures. The Cryo-Tags can also combat most organic solvents and caustic agents. Compatible with most surfaces, like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in a pack of 1200 labels in the colors of blue, green, orange, red, white and yellow.
Catalog No. NC9630776