Learn More
Spherotech Rainbow Fluorescent Particle Slide, 6 Sizes (0.56, 0.9, 3.1, 5.5, 9.9, & 16.2 um)
Supplier:  Spherotech FPSM576
Encompass_Preferred
The Fluorescent Particle Slides are prepared by mounting fluorescent particles with single or multiple fluorophores in a permanent mounting medium. The Fluorescent Particle Slides are very convenient to use and can be used for routine alignment and calibration of confocal fluorescent microscopes, laser scanning cytometers and other image systems.
Catalog No. NC1559715