Learn More
Description
- Measurement and fractions are commonly cited as 2 of the hardest concepts to teach in math.
- Our Rainbow Fraction Measuring Cups provide a unique 3D representation that helps connect volume and fractions.
- Children see and experience measuring, comparing volumes and capacity, and learning fractional relationships.
- For over 50 years, hand2mind has encouraged hands-on learning and discovery, driving deeper understanding and helping children unlock their full potential. Hand2mind specializes in math manipulatives designed to help students learn by doing.
- Particularly with fractions, our rainbow fraction tiles, rainbow fraction circles and rainbow fraction tower cubes make learning fractions as easy as learning the colors of the rainbow. From early math to middle school math, hand2mind has you covered with products designed to enhance learning in the classroom and the world around us.
- Through hands-on exploration and discovery we strive to find new ways to encourage students in math practice, literacy activities, STEM education and more!
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 8 to 10 Students |
| Product Type | Fractions, Decimals, Percents |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
