Learn More
Med Vet International Rainbow Jacquette 34-35, Rainbow Yellow, J0973DN, Each
Catalog No. 5000583935
Encompass_Preferred
Universal scaler: sickle scaler for supragingival on posterior surfaces. Part of the Small Animal Prophy Kit (J0973n).
Catalog No. 50-005-83935 Supplier Med Vet International Supplier No. JOR973DN
Universal scaler: sickle scaler for supragingival on posterior surfaces. Part of the Small Animal Prophy Kit (J0973n).