Learn More
Cole-Parmer Write-On Tape, Assorted Colors, 3/4" x 14 yds, 16/pk
Supplier:  Cole-Parmer 0653038
Encompass_Preferred
Cole-Parmer Write-On Tape, Assorted Colors, 3/4" x 14 yds, 16/pk.Use tape to identify specific applications by color coding. Durable labeling tape is waterproof, and oil and acid resistant. Tape can be used with pencil, pen, or permanent marker. Withstands temperatures from 10 to 250F (23 to 121C). *Note Tape Dispenser Rack Note Included
Catalog No. NC0647069