Learn More
MSC Rainbow Technology Desiccant, Silica Gel, 416 g, Mil-D-3464, 5 Pack
Catalog No. 5000793026
Encompass
This activated silica gel desiccant is designed for bagged packaging use and static dehumidification. Each 416 gram pack conforms to Mil-D-3464 Type I & II, providing effective moisture removal.
  • Composed of silica gel
  • Conforms to Mil-D-3464 Type I & II
  • Ideal for packaging and static dehumidification applications
  • Reactivation temperature in bag: 245°F
  • Minimum reactivation time: 16 hours
  • Packaged as 5 packs per bag
  • Individual pack weight: 416 grams
Catalog No. 50-007-93026 Supplier MSC Supplier No. 87845152
This activated silica gel desiccant is designed for bagged packaging use and static dehumidification. Each 416 gram pack conforms to Mil-D-3464 Type I & II, providing effective moisture removal.
  • Composed of silica gel
  • Conforms to Mil-D-3464 Type I & II
  • Ideal for packaging and static dehumidification applications
  • Reactivation temperature in bag: 245°F
  • Minimum reactivation time: 16 hours
  • Packaged as 5 packs per bag
  • Individual pack weight: 416 grams