Learn More
PHOENIX SCIENTIFIC RAINBOW TROUT SERUM 50ML
Catalog No. NC3002342
Encompass_Preferred
  • High-quality biological reagent
  • Sourced from healthy, pathogen-free Rainbow Trout
  • Minimizes cross-reactivity with mammalian sources
  • Supports cell culture, immunoassay development
  • Available in a 50 mL liquid format
  • Filtration at 0.22 µm sterile-filtered
  • Long-term storage at -20°C
  • Short-term storage option of 2–8°C
  • Ideal alternative to traditional sera
  • Applicable for aquatic health management studies
Catalog No. NC3002342 Supplier PHOENIX SCIENTIFIC Supplier No. FS303
  • High-quality biological reagent
  • Sourced from healthy, pathogen-free Rainbow Trout
  • Minimizes cross-reactivity with mammalian sources
  • Supports cell culture, immunoassay development
  • Available in a 50 mL liquid format
  • Filtration at 0.22 µm sterile-filtered
  • Long-term storage at -20°C
  • Short-term storage option of 2–8°C
  • Ideal alternative to traditional sera
  • Applicable for aquatic health management studies