Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ XTIP™ Rainin LTS Compatible TipEject Reload

Catalog No. 12111428 Shop All OHAUS Products
Change view
Click to view available options
Volume (Metric):
20 μL
250 μL
300 μL
1000 μL
1200 μL
Color Code:
Blue
Gray
Green
Red
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Code
12-111-428 300 μL Green
12-111-386 20 μL Red
12-111-387 250 μL Green
12-111-389 1000 μL Blue
12-111-390 1200 μL Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12-111-428 Supplier BIOTIX™ Supplier No. 63305951
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BIOTIX™ Rainin LTS Compatible TipEject Reload creates an ergonomic system in the lab with FlexFit™, StarStop™ and an LTS™ style pipette. Manual ejection racking

  • FlexFit™ technology creates a seal with less insertion forces than that of competitors
  • StarStop™ works with the Rainin LTS pipette to eliminate high ejection forces
  • X-Resin™ maximizes sample recovery and accuracy by reducing surface to sample binding
  • Blade™ reduces the frequency of hanging droplet formation, improving precision and accuracy
  • Tip Eject saves space on lab bench while providing an easy manual ejection system for racking tips

Order Info

960 tips/pack

Certifications

ISO 9001:2015, Certified free of DNASE, RNASE, Endotoxins/pyrogens, PCR inhibitors. Sterilized product is certified to 10^-3 SAL.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 300 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Rack Color Clear
Autoclavable Autoclavable
Length (Metric) 58.93 mm
Clean Claims RNase, DNase, ATP, pyrogen, endotoxin and DNA Free
Sterilization Method E-Beam
Features FlexFit, StarStop, X-Resin, Blade
Format Refill
Tip Style Standard, Ergonomic, Low Retention
Color Code Green
Disposable Single-use
For Use With (Equipment) Rainin LTS
Graduated Yes
Product Line xTIP
Material Polypropylene
Fit for Purpose Certified Quality
Graduations 10 μL, 50 μL, 100 μL, 200 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.