Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Great Lakes Diagnostics Rainin LTS compatible tips

Catalog No. 23444016
Change view
Click to view available options
Volume (Metric):
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
23-444-016 20 μL
23-444-017 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-444-016 Supplier Great Lakes Diagnostics Supplier No. ASPMB20RFS9
Add to Cart
Add to Cart
This item is not returnable. View return policy

Readily available Rainin LTS compatible pipette tips. Available in sterile, filtered convenient packaging.

  • Hanging rack containg 96 tips
  • 4800/Cs
  • Polyproplyne
  • Sterile
  • Filtered
  • Certified DNase/RNase free
  • Compatible with Rainin LTS pipettors and work stations

Specifications

Volume (Metric) 20 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Length (Metric) 49.45 mm
Features Certified DNase/RNase free
Format Hinged Rack Sterile
Tip Style Automation
Quantity 4800/Cs.
Disposable Single-use
For Use With (Equipment) Rainin LTS
Graduated Yes
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.