Learn More
Pipette Supplies, Inc. Rainin Pipet-Lite Seal, Single Channel, 2μL (Pipette Supplies)
SDP
Catalog No. NC1908821
Encompass
Piston seal for the following Rainin 2μL Pipettes:Pipet-Lite (L, SL)Pipet-Lite XLS (XLS)Pipet-Plus (R, RL)EDP1 (E1, SE1)EDP3, EDP3 Plus (E3, SE3)E-Man Hybrid (H, SH)Alternative to Rainin 6200-131 / 17004643
Catalog No. NC1908821 Supplier Pipette Supplies, Inc. Supplier No. 80002/5
Piston seal for the following Rainin 2μL Pipettes:Pipet-Lite (L, SL)Pipet-Lite XLS (XLS)Pipet-Plus (R, RL)EDP1 (E1, SE1)EDP3, EDP3 Plus (E3, SE3)E-Man Hybrid (H, SH)Alternative to Rainin 6200-131 / 17004643