Learn More
National Safety Apparel™ Rainwear Bib Pants Polyurethane Coated FR Weft
SDP
Catalog No. 170012181 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2181 Small
17-001-2182 Medium
17-001-2183 Large
17-001-2184 X-Large
17-001-2185 2X-Large
17-001-2186 3X-Large
17-001-2187 4X-Large
17-001-2188 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-2181 Supplier National Safety Apparel™ Supplier No. R40RL19SM
Add to Cart
Add to Cart
Specifications
Closure Type Buckle
Color Fluorescent Yellow
Material Polyurethane-coated FR Cotton
Seam Type Welded
Size Small
Suspender Adjustable
Ankle Style Open
Arc Flash Rating 19 cal/cm² (Fluorescent Yellow); 24 cal/cm² (Fluorescent Orange)
Certifications/Compliance CAT 2, NFPA 70 E, ANSI/ISEA 107 Class E, CSA Z462, ASTM F2733, ASTM F1891, OSHA 1910.269
Disposable Reusable