Learn More
Research Products International Corp Raised Rim PCR Plate, 250 ul, Compatible with Corning, Natural, 50 per Package
SDP
Supplier:  Research Products International Corp 141344
Encompass
Rigid 96 well plates feature 8 x 12 format designed for high performance thermal cycling and automation applications. Each well makes full contact with the heat block. Consistent heat transfer is ensured by a uniform wall thickness. Special polypropylene material provides a non-wettable surface for easy sample recovery.
Plates can be cut in strips of 16 wells (2 x 8 format).
Catalog No. 50-197-7463