Learn More
Siemens RAL Diff Quick
Catalog No. NC1433921
Encompass_Preferred
RAL Diff-Quik is a simplified and cost-effective alternative to Wright-Giemsa staining and is faster than typical staining procedures. It reduces the process from 4 minutes to 15 seconds. The speed of execution makes it an ideal stain for emergency diagnostics of medullary and blood smears.
This item is not returnable. View return policy
Catalog No. NC1433921 Supplier Siemens Supplier No. 10736133
RAL Diff-Quik is a simplified and cost-effective alternative to Wright-Giemsa staining and is faster than typical staining procedures. It reduces the process from 4 minutes to 15 seconds. The speed of execution makes it an ideal stain for emergency diagnostics of medullary and blood smears.