Learn More
Aviva Systems Biology Corp RALA Antibody - middle region (ARP56643_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56643_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RALA. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RALA. Alternate names: MGC48949, RALApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-184