Learn More
ABclonal Technology RALA Rabbit pAb
SDP
Supplier:  ABclonal Technology A1173620UL
Encompass_Preferred
The product of this gene belongs to the small GTPase superfamily, Ras family of proteins. GTP-binding proteins mediate the transmembrane signaling initiated by the occupancy of certain cell surface receptors. This gene encodes a low molecular mass ras-like GTP-binding protein that shares about 50% similarity with other ras proteins.
Catalog No. 50-001-16222