Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ralaniten triacetate | 1637573-04-6 | 98.8% | 521.00 | C27H33ClO8 | 10MM 1ML
SDP

Catalog No. 5000238820
Encompass_Preferred

Ralaniten triacetate (EPI-506) is an androgen receptor N-terminal domain (AR NTD) inhibitor supplied for research use. It is offered as a ready-to-use 10 mM solution in DMSO and in multiple solid mass pack sizes, with analytical purity suitable for in vitro studies. The compound is intended for preclinical research applications where AR NTD modulation is required.

  • Targets the androgen receptor N-terminal domain.
  • Available as a 10 mM solution in DMSO and in various solid sizes.
  • High purity (~98.8%) for reliable experimental results.
  • Molecular formula C27H33ClO8 and molecular weight 521.00 for reference.
  • Recommended storage: solid at -20°C, solution at -80°C for long-term storage.
  • Identified by CAS 1637573-04-6 for unambiguous reference.

Catalog No. 50-002-38820 Supplier Medchemexpress LLC Supplier No. HY123875A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ralaniten triacetate (EPI-506) is an androgen receptor N-terminal domain (AR NTD) inhibitor supplied for research use. It is offered as a ready-to-use 10 mM solution in DMSO and in multiple solid mass pack sizes, with analytical purity suitable for in vitro studies. The compound is intended for preclinical research applications where AR NTD modulation is required.

  • Targets the androgen receptor N-terminal domain.
  • Available as a 10 mM solution in DMSO and in various solid sizes.
  • High purity (~98.8%) for reliable experimental results.
  • Molecular formula C27H33ClO8 and molecular weight 521.00 for reference.
  • Recommended storage: solid at -20°C, solution at -80°C for long-term storage.
  • Identified by CAS 1637573-04-6 for unambiguous reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.