Learn More
Medchemexpress LLC Ralaniten triacetate | 1637573-04-6 | 98.8% | 521.00 | C27H33ClO8 | 25MG
SDP
Catalog No. 5000236012
Encompass_Preferred
Ralaniten triacetate is an orally active androgen receptor (AR) N-terminal domain (NTD) inhibitor used for preclinical research. It functions as a pro-agent of ralaniten, inhibiting AR transcriptional activity and showing activity against full-length and resistance-related AR species, including AR-v7. Consult the supplier documentation for handling, storage, purity, and safety information.
  • First-in-class AR N-terminal domain inhibition.
  • Active against full-length and resistant AR species such as AR-v7.
  • Inhibits AR transcriptional activity.
  • Suitable for research into castration-resistant prostate cancer models.
  • Supplied with certificate of analysis and safety data sheet for quality verification.
Catalog No. 50-002-36012 Supplier Medchemexpress LLC Supplier No. HY123875A25MG
Ralaniten triacetate is an orally active androgen receptor (AR) N-terminal domain (NTD) inhibitor used for preclinical research. It functions as a pro-agent of ralaniten, inhibiting AR transcriptional activity and showing activity against full-length and resistance-related AR species, including AR-v7. Consult the supplier documentation for handling, storage, purity, and safety information.
  • First-in-class AR N-terminal domain inhibition.
  • Active against full-length and resistant AR species such as AR-v7.
  • Inhibits AR transcriptional activity.
  • Suitable for research into castration-resistant prostate cancer models.
  • Supplied with certificate of analysis and safety data sheet for quality verification.