Learn More
Medchemexpress LLC Ralaniten triacetate | 1637573-04-6 | 98.8% | 521.00 g/mol | C27H33ClO8 | 1MG
SDP
Catalog No. 5000237978
Encompass_Preferred
Ralaniten triacetate (EPI-506) is a pro-drug of ralaniten and an orally active small-molecule inhibitor of the androgen receptor N-terminal domain (NTD) used in preclinical research on castration-resistant prostate cancer. It inhibits AR transcriptional activity, including resistance-related AR species such as AR-v7, and is supplied for laboratory research use only.
  • Inhibits androgen receptor N-terminal domain activity.
  • Active against full-length and variant AR species, including AR-v7.
  • High purity suitable for research (98.79%).
  • Available in small milligram quantities and as DMSO solutions for assay use.
  • Chemical formula C27H33ClO8; molecular weight 521.00 g/mol.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-37978 Supplier Medchemexpress LLC Supplier No. HY123875A1MG
Ralaniten triacetate (EPI-506) is a pro-drug of ralaniten and an orally active small-molecule inhibitor of the androgen receptor N-terminal domain (NTD) used in preclinical research on castration-resistant prostate cancer. It inhibits AR transcriptional activity, including resistance-related AR species such as AR-v7, and is supplied for laboratory research use only.
  • Inhibits androgen receptor N-terminal domain activity.
  • Active against full-length and variant AR species, including AR-v7.
  • High purity suitable for research (98.79%).
  • Available in small milligram quantities and as DMSO solutions for assay use.
  • Chemical formula C27H33ClO8; molecular weight 521.00 g/mol.
  • For research use only; not for human or veterinary use.