Learn More
Medchemexpress LLC HY-108841 50mg , Raleukin CAS:143090-92-0 Purity:>98%
SDP
Catalog No. 502259124
Encompass_Preferred
HY-108841 50mg Raleukin CAS: 143090-92-0 Raleukin (AMG-719) is a recombinant, nonglycosylated human interleukin-1 receptor (IL-1R) antagonist. Raleukin (AMG-719) is the first biological agent to block the pro-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9124 Supplier Medchemexpress LLC Supplier No. HY10884150MG
HY-108841 50mg Raleukin CAS: 143090-92-0 Raleukin (AMG-719) is a recombinant, nonglycosylated human interleukin-1 receptor (IL-1R) antagonist. Raleukin (AMG-719) is the first biological agent to block the pro-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.