Learn More
Medchemexpress LLC HY-101437 10mg Medchemexpress, Ralfinamide CAS:133865-88-0 Purity:>98%
SDP
Catalog No. 501932129
Encompass_Preferred
Medchemexpress, HY-101437 10mg Ralfinamide CAS:133865-88-0 Ralfinamide (FCE-26742A) is an orally available Na + blocker derived from α-aminoamide, with function of suppressing pain [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2129 Supplier Medchemexpress LLC Supplier No. HY10143710MG
Medchemexpress, HY-101437 10mg Ralfinamide CAS:133865-88-0 Ralfinamide (FCE-26742A) is an orally available Na + blocker derived from α-aminoamide, with function of suppressing pain [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.