Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED ON-TARGETplus Human RALGAPA2 (57186) siRNA - SMARTpool, Unit Size: 5 nmol
Catalog No. NC3240894
Encompass
Knockdown your target gene function with confidenceThe patented modification pattern for specificity, combined with the SMARTselection algorithm for efficient, guaranteed target gene silencing, makes ON-TARGETplus siRNA the premium choice for optimal knockdown and reduced off-targets.Available as pre-designed, genome-wide reagents for human, mouse and rat.Off-targets reduced by up to 90% compared to unmodified siRNAGuaranteed silencing by SMARTpool and 3 of 4 individual siRNAs (see Guarantee tab)Sequence information provided with siRNA purchase
Catalog No. NC3240894 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. L024244010005
Add to Cart
Add to Cart
Knockdown your target gene function with confidenceThe patented modification pattern for specificity, combined with the SMARTselection algorithm for efficient, guaranteed target gene silencing, makes ON-TARGETplus siRNA the premium choice for optimal knockdown and reduced off-targets.Available as pre-designed, genome-wide reagents for human, mouse and rat.Off-targets reduced by up to 90% compared to unmodified siRNAGuaranteed silencing by SMARTpool and 3 of 4 individual siRNAs (see Guarantee tab)Sequence information provided with siRNA purchase