Learn More
Aviva Systems Biology Corp RALGPS1 Antibody - middle region (ARP42415_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42415_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RALGPS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RALGPS1. Alternate names: KIAA0351, RALGEF2, RALGPS1A, RP13-225O21.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58219