Learn More
Aviva Systems Biology Corp RALGPS2 Antibody - C-terminal region (ARP57092_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57092_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RALGPS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RALGPS2. Alternate names: RP4-595C2.1, FLJ10244, FLJ25604, KIAA0351, dJ595C2.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62076