Learn More
TARGETMOL CHEMICALS INC RALIMETINIB DIMESYLATE 10MG
SDP
Catalog No. 502576617
Encompass_Preferred
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ralimetinib dimesylate (LY2228820 dimesylate) is the dimesylate salt form of LY2228820 a tri-substituted imidazole derivative and orally available p38 mitogen-activated protein kinase (MAPK) inhibitor with potential anti-inflammatory and antineoplastic activities. purity: 99%
Catalog No. 50-257-6617 Supplier TARGETMOL CHEMICALS INC Supplier No. T604710MG
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ralimetinib dimesylate (LY2228820 dimesylate) is the dimesylate salt form of LY2228820 a tri-substituted imidazole derivative and orally available p38 mitogen-activated protein kinase (MAPK) inhibitor with potential anti-inflammatory and antineoplastic activities. purity: 99%