Learn More
Cayman Chemical RalmItaront 10mg
SDP
Catalog No. 502978487
Encompass_Preferred
A TAAR1 agonist; induces cAMP accumulation in HEK293T cells expressing human TAAR1 (EC50 = 17.78 nM)
Catalog No. 50-297-8487 Supplier Cayman Chemical Supplier No. 4111110 MG
A TAAR1 agonist; induces cAMP accumulation in HEK293T cells expressing human TAAR1 (EC50 = 17.78 nM)