Learn More
Cayman Chemical Raloxifene-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 1188263-47-9, 1 mg
SDP
Catalog No. 502994232
Encompass_Preferred
An internal standard for the quantification of raloxifene by GC- or LC-MS
Catalog No. 50-299-4232 Supplier Cayman Chemical Supplier No. 229001 MG
An internal standard for the quantification of raloxifene by GC- or LC-MS