Learn More
Medchemexpress LLC Raloxifene-d4 hydrochloride | 1188263-47-9 | 98.6% | 514.07 | 500 UG
SDP
Catalog No. 5000351315
Encompass_Preferred
Raloxifene-d4 hydrochloride is the deuterium labeled form of Raloxifene, a benzothiophene-derived selective estrogen receptor modulator (SERM). It exhibits estrogen-agonistic effects on bone and lipids, and estrogen-antagonistic effects on the breast and uterus. This compound is utilized in breast cancer and osteoporosis research.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits estrogen-agonistic effects on bone and lipids
  • Shows estrogen-antagonistic effects on the breast and uterus
  • Valuable for breast cancer and osteoporosis research
Catalog No. 50-003-51315 Supplier Medchemexpress LLC Supplier No. HY13738S2500UG
Raloxifene-d4 hydrochloride is the deuterium labeled form of Raloxifene, a benzothiophene-derived selective estrogen receptor modulator (SERM). It exhibits estrogen-agonistic effects on bone and lipids, and estrogen-antagonistic effects on the breast and uterus. This compound is utilized in breast cancer and osteoporosis research.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits estrogen-agonistic effects on bone and lipids
  • Shows estrogen-antagonistic effects on the breast and uterus
  • Valuable for breast cancer and osteoporosis research