Learn More
Selleck Chemical LLC Raloxifene HCl, 50mg | CAS No. 82640-04-8 | Purity: 99.99%
SDP
Catalog No. NC3662312
Encompass_Preferred
Raloxifene (LY156758, Keoxifene) HCl is a selective and orally active estrogen receptor modulator (SERM), which inhibits human cytosolic aldehyde oxidase-catalyzed phthalazine oxidation activity with IC50 of 5.7 nM.
Catalog No. NC3662312 Supplier Selleck Chemical LLC Supplier No. S122750MG
Raloxifene (LY156758, Keoxifene) HCl is a selective and orally active estrogen receptor modulator (SERM), which inhibits human cytosolic aldehyde oxidase-catalyzed phthalazine oxidation activity with IC50 of 5.7 nM.