Learn More
Medchemexpress LLC HY-13738A 100mg , Raloxifene (hydrochloride) LY156758 hydrochloride CAS:82640-04-8 Purity:98%
SDP
Catalog No. 501493031
Encompass_Preferred
Medchemexpress, HY-13738A 100mg Raloxifene (hydrochloride) LY156758 hydrochloride CAS:82640-04-8 Formula:C28H28ClNO4S Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3031 Supplier Medchemexpress LLC Supplier No. HY13738A100MG
Medchemexpress, HY-13738A 100mg Raloxifene (hydrochloride) LY156758 hydrochloride CAS:82640-04-8 Formula:C28H28ClNO4S Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.