Learn More
Medchemexpress LLC Raloxifene (hydrochloride) | 82640-04-8 | 99.8% | 510.04 | 1 ML
SDP
Catalog No. 5000349732
Encompass_Preferred
Raloxifene hydrochloride is a second-generation selective and orally active estrogen receptor modulator. It exhibits estrogen-agonistic effects on bone and lipid metabolism and estrogen-antagonistic effects on uterine endometrium and breast tissue.
  • Estrogen-agonistic effects on bone and lipid metabolism
  • Estrogen-antagonistic effects on uterine endometrium and breast tissue
  • For research use only
  • Purity of 99.8%
  • Stable under recommended storage conditions
Catalog No. 50-003-49732 Supplier Medchemexpress LLC Supplier No. HY13738A10MM/1ML
Raloxifene hydrochloride is a second-generation selective and orally active estrogen receptor modulator. It exhibits estrogen-agonistic effects on bone and lipid metabolism and estrogen-antagonistic effects on uterine endometrium and breast tissue.
  • Estrogen-agonistic effects on bone and lipid metabolism
  • Estrogen-antagonistic effects on uterine endometrium and breast tissue
  • For research use only
  • Purity of 99.8%
  • Stable under recommended storage conditions