Learn More
Medchemexpress LLC Raloxifene (hydrochloride) | 82640-04-8 | 99.8% | 510.04 | 100 MG
SDP
Catalog No. 5000348571
Encompass_Preferred
Raloxifene hydrochloride is a second generation selective and orally active estrogen receptor modulator. It produces estrogen-agonistic effects on bone and lipid metabolism and estrogen-antagonistic effects on uterine endometrium and breast tissue. It is for research use only and not sold to patients.
  • Second generation selective and orally active estrogen receptor modulator
  • Estrogen-agonistic effects on bone and lipid metabolism
  • Estrogen-antagonistic effects on uterine endometrium and breast tissue
  • For research use only
Catalog No. 50-003-48571 Supplier Medchemexpress LLC Supplier No. HY13738AR100MG
Raloxifene hydrochloride is a second generation selective and orally active estrogen receptor modulator. It produces estrogen-agonistic effects on bone and lipid metabolism and estrogen-antagonistic effects on uterine endometrium and breast tissue. It is for research use only and not sold to patients.
  • Second generation selective and orally active estrogen receptor modulator
  • Estrogen-agonistic effects on bone and lipid metabolism
  • Estrogen-antagonistic effects on uterine endometrium and breast tissue
  • For research use only